Assay Details
Gene Symbol: prdx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030437.1 | NP_001025608.1 | 281 | 84 | ||
GenBank mRNA | BC091062.1 | - | 281 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
