Assay Details
Gene Symbol: tha1p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030440.1 | NP_001025611.1 | 2 - 3 | 241 | 99 | |
XM_012966670.2 | XP_012822124.1 | 2 - 3 | 316 | 99 | ||
XM_012966671.1 | XP_012822125.1 | 2 - 3 | 421 | 99 | ||
GenBank mRNA | BC091069.1 | - | 2 - 3 | 241 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
