Assay Details
Gene Symbol: ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030452.1 | NP_001025623.1 | 10 - 11 | 1168 | 76 | |
XM_012966672.1 | XP_012822126.1 | 10 - 11 | 1211 | 76 | ||
GenBank mRNA | BC091090.1 | - | 10 - 11 | 1168 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
