Assay Details
Gene Symbol: mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030492.1 | NP_001025663.1 | 6 - 7 | 608 | 92 | |
GenBank mRNA | BC092545.1 | - | 6 - 7 | 608 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
