Assay Details
| Gene Symbol: scd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032329.1 | NP_001027500.1 | 2 - 3 | 275 | 73 | |
| XM_012966676.2 | XP_012822130.1 | 2 - 3 | 703 | 73 | ||
| GenBank mRNA | BC096632.1 | - | 2 - 3 | 275 | 73 | |
More Information
Set Membership: |
|

scd
Genomic Map