Assay Details
| Gene Symbol: cox7a2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032330.1 | NP_001027501.1 | 1 - 2 | 102 | 100 | |
| XM_018093924.1 | XP_017949413.1 | 1 - 2 | 100 | |||
| GenBank mRNA | BC096633.1 | - | 1 - 2 | 102 | 100 | |
| CT025416.2 | - | 1 - 2 | 116 | 100 | ||
More Information
Set Membership: |
|

cox7a2l
Genomic Map