Assay Details
Gene Symbol: ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032334.1 | NP_001027505.1 | 1 - 2 | 133 | 59 | |
XM_012959155.2 | XP_012814609.1 | 1 - 2 | 304 | 59 | ||
XM_018092127.1 | XP_017947616.1 | 1 - 2 | 305 | 59 | ||
GenBank mRNA | BC098082.1 | - | 1 - 2 | 133 | 59 | |
CU075829.1 | - | 1 - 2 | 101 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
