Assay Details
| Gene Symbol: marcks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006692.2 | NP_001006693.2 | 2 - 3 | 280 | 74 | |
| GenBank mRNA | BC075394.1 | - | 1 - 2 | 572 | 74 | |
| CR848092.2 | - | 2 - 3 | 280 | 74 | ||
More Information
Set Membership: |
|

marcks
Genomic Map