Assay Details
Gene Symbol: arf6.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006796.2 | NP_001006797.1 | 1 | 853 | 86 | |
GenBank mRNA | BC076664.1 | - | 1 | 681 | 86 | |
CR762323.2 | - | 1 | 853 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
