Assay Details
Gene Symbol: pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007507.3 | NP_001007508.2 | 6 - 7 | 896 | 99 | |
XM_012960904.2 | XP_012816358.1 | 6 - 7 | 911 | 99 | ||
XM_012960906.2 | XP_012816360.1 | 4 - 5 | 549 | 99 | ||
XM_018093011.1 | XP_017948500.1 | 6 - 7 | 930 | 99 | ||
XM_018093012.1 | XP_017948501.1 | 6 - 7 | 930 | 99 | ||
GenBank mRNA | BC079944.1 | - | 6 - 7 | 896 | 99 | |
CR848635.2 | - | 4 - 5 | 510 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
