Assay Details
| Gene Symbol: slc25a44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008080.2 | NP_001008081.1 | 1 - 2 | 721 | 104 | |
| GenBank mRNA | BC080980.1 | - | 1 - 2 | 840 | 104 | |
| CR855404.2 | - | 1 - 2 | 721 | 104 | ||
More Information
Set Membership: |
|

slc25a44
Genomic Map