Assay Details
| Gene Symbol: prps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011074.2 | NP_001011074.1 | 2 - 3 | 497 | 62 | |
| XM_012956842.2 | XP_012812296.1 | 2 - 3 | 901 | 62 | ||
| GenBank mRNA | BC084459.1 | - | 2 - 3 | 440 | 62 | |
| CR848150.2 | - | 2 - 3 | 497 | 62 | ||
More Information
Set Membership: |
|

prps2
Genomic Map