Assay Details
| Gene Symbol: tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011324.2 | NP_001011324.1 | 3 - 4 | 535 | 70 | |
| GenBank mRNA | BC088075.1 | - | 3 - 4 | 421 | 70 | |
| CR761327.2 | - | 3 - 4 | 535 | 70 | ||
More Information
Set Membership: |
|

tm2d2
Genomic Map