Assay Details
Gene Symbol: rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011394.2 | NP_001011394.2 | 6 - 7 | 823 | 76 | |
XM_018091215.1 | XP_017946704.1 | 6 - 7 | 936 | 76 | ||
GenBank mRNA | BC088588.1 | - | 6 - 7 | 809 | 76 | |
CR760337.2 | - | 6 - 7 | 823 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
