Assay Details
Gene Symbol: rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011394.2 | NP_001011394.2 | 10 - 11 | 1168 | 69 | |
XM_018091215.1 | XP_017946704.1 | 10 - 11 | 1284 | 69 | ||
GenBank mRNA | BC088588.1 | - | 10 - 11 | 1154 | 69 | |
CR760337.2 | - | 10 - 11 | 1168 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
