Assay Details
| Gene Symbol: rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011394.2 | NP_001011394.2 | 11 - 12 | 1278 | 91 | |
| XM_018091215.1 | XP_017946704.1 | 11 - 12 | 1394 | 91 | ||
| GenBank mRNA | BC088588.1 | - | 11 - 12 | 1264 | 91 | |
| CR760337.2 | - | 11 - 12 | 1278 | 91 | ||
More Information
Set Membership: |
|

rbbp4
Genomic Map