Assay Details
Gene Symbol: wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011411.2 | NP_001011411.1 | 3 - 4 | 320 | 71 | |
GenBank mRNA | BC067938.1 | - | 3 - 4 | 383 | 72 | |
BC088786.1 | - | 3 - 4 | 424 | 71 | ||
CR761006.2 | - | 3 - 4 | 320 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
