Assay Details
| Gene Symbol: urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011486.2 | NP_001011486.2 | 3 - 4 | 337 | 66 | |
| GenBank mRNA | CR760078.2 | - | 3 - 4 | 337 | 66 | |
More Information
Set Membership: |
|

urod
Genomic Map