Assay Details
| Gene Symbol: rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015915.2 | NP_001015915.1 | 3 - 4 | 432 | 81 | |
| GenBank mRNA | BC170836.1 | - | 3 - 4 | 419 | 81 | |
| BC170838.1 | - | 3 - 4 | 419 | 81 | ||
| CR926418.2 | - | 3 - 4 | 432 | 81 | ||
More Information
Set Membership: |
|

rala
Genomic Map