Assay Details
| Gene Symbol: gosr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015945.2 | NP_001015945.1 | 3 - 4 | 259 | 66 | |
| XM_012956914.2 | XP_012812368.1 | 3 - 4 | 339 | 66 | ||
| GenBank mRNA | BC098325.1 | - | 3 - 4 | 229 | 66 | |
| BC161527.1 | - | 3 - 4 | 310 | 66 | ||
| CR848485.2 | - | 3 - 4 | 259 | 66 | ||
More Information
Set Membership: |
|

gosr1
Genomic Map