Assay Details
| Gene Symbol: chst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015949.2 | NP_001015949.1 | 1 | 1244 | 84 | |
| XM_012970394.2 | XP_012825848.1 | 2 | 1341 | 84 | ||
| XM_012970395.2 | XP_012825849.1 | 3 | 1427 | 84 | ||
| XM_012970396.2 | XP_012825850.1 | 4 | 1463 | 84 | ||
| XM_012970397.2 | XP_012825851.1 | 2 | 1473 | 84 | ||
| XM_012970398.2 | XP_012825852.1 | 3 | 1562 | 84 | ||
| GenBank mRNA | BC123055.1 | - | 1 | 1215 | 84 | |
| CR848482.2 | - | 1 | 1244 | 84 | ||
More Information
Set Membership: |
|

chst12
Genomic Map