Assay Details
Gene Symbol: tpte2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015951.2 | NP_001015951.1 | 15 - 16 | 1297 | 112 | |
XM_018091231.1 | XP_017946720.1 | 15 - 16 | 1555 | 112 | ||
XM_018091232.1 | XP_017946721.1 | 16 - 17 | 1619 | 112 | ||
XM_018091233.1 | XP_017946722.1 | 16 - 17 | 2192 | 112 | ||
XM_018091234.1 | XP_017946723.1 | 15 - 16 | 1270 | 112 | ||
GenBank mRNA | BC121583.1 | - | 10 - 11 | 768 | 112 | |
BC154861.1 | - | 12 - 13 | 1037 | 112 | ||
CR848475.2 | - | 15 - 16 | 1297 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
