Assay Details
| Gene Symbol: egln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015960.2 | NP_001015960.1 | 2 - 3 | 1059 | 105 | |
| GenBank mRNA | BC161035.1 | - | 2 - 3 | 1175 | 105 | |
| BC170673.1 | - | 2 - 3 | 1019 | 105 | ||
| BC170677.1 | - | 2 - 3 | 1019 | 105 | ||
| CR848438.2 | - | 2 - 3 | 1059 | 105 | ||
More Information
Set Membership: |
|

egln1
Genomic Map