Assay Details
Gene Symbol: frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012969325.2 | XP_012824779.1 | 6 - 7 | 917 | 94 | |
XM_012969326.2 | XP_012824780.1 | 6 - 7 | 918 | 94 | ||
XM_012969327.2 | XP_012824781.1 | 6 - 7 | 917 | 94 | ||
XM_018096717.1 | XP_017952206.1 | 6 - 7 | 914 | 94 | ||
XM_018096718.1 | XP_017952207.1 | 3 - 4 | 351 | 94 | ||
GenBank mRNA | CR848147.2 | - | 6 - 7 | 884 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
