Assay Details
Gene Symbol: c18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016028.2 | NP_001016028.1 | 2 - 3 | 847 | 73 | |
GenBank mRNA | BC171315.1 | - | 2 - 3 | 823 | 73 | |
CR848138.2 | - | 2 - 3 | 847 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
