Assay Details
Gene Symbol: gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016037.2 | NP_001016037.1 | 1 - 2 | 162 | 77 | |
XM_012962962.2 | XP_012818416.1 | 2 - 3 | 766 | 77 | ||
XM_012962963.2 | XP_012818417.1 | 2 - 3 | 966 | 77 | ||
GenBank mRNA | BC157274.1 | - | 2 - 3 | 362 | 77 | |
CR848114.2 | - | 1 - 2 | 162 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
