Assay Details
| Gene Symbol: rad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016045.2 | NP_001016045.1 | 2 - 3 | 568 | 75 | |
| XM_012961937.2 | XP_012817391.1 | 3 - 4 | 388 | 75 | ||
| XM_012961942.2 | XP_012817396.1 | 2 - 3 | 577 | 75 | ||
| GenBank mRNA | BC157217.1 | - | 2 - 3 | 495 | 75 | |
| CR762361.2 | - | 2 - 3 | 568 | 75 | ||
More Information
Set Membership: |
|

rad1
Genomic Map