Assay Details
Gene Symbol: taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016066.2 | NP_001016066.1 | 3 - 4 | 210 | 73 | |
GenBank mRNA | CR762262.2 | - | 3 - 4 | 210 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
