Assay Details
| Gene Symbol: akirin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016080.2 | NP_001016080.1 | 4 - 5 | 648 | 66 | |
| GenBank mRNA | BC090594.1 | - | 4 - 5 | 677 | 66 | |
| CR762217.2 | - | 4 - 5 | 648 | 66 | ||
More Information
Set Membership: |
|

akirin1
Genomic Map