Assay Details
Gene Symbol: rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016091.2 | NP_001016091.1 | 3 - 4 | 332 | 57 | |
GenBank mRNA | BC171269.1 | - | 3 - 4 | 321 | 57 | |
BC171271.1 | - | 3 - 4 | 321 | 57 | ||
CR762181.2 | - | 3 - 4 | 332 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
