Assay Details
| Gene Symbol: rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016096.2 | NP_001016096.1 | 261 | 76 | ||
| GenBank mRNA | BC135723.1 | - | 284 | 76 | ||
| CR762159.2 | - | 261 | 76 | |||
More Information
Set Membership: |
|

rbm7
Genomic Map