Assay Details
| Gene Symbol: lbhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016100.2 | NP_001016100.1 | 2 - 3 | 173 | 109 | |
| GenBank mRNA | BC161504.1 | - | 2 - 3 | 250 | 109 | |
| BC166939.1 | - | 2 - 3 | 200 | 109 | ||
| CR762146.2 | - | 2 - 3 | 173 | 109 | ||
More Information
Set Membership: |
|

lbhd1
Genomic Map