Assay Details
Gene Symbol: ccndx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016108.2 | NP_001016108.1 | 1 - 2 | 237 | 69 | |
GenBank mRNA | BC158907.1 | - | 1 - 2 | 240 | 69 | |
CR762119.2 | - | 1 - 2 | 237 | 69 | ||
DQ508541.1 | - | 1 - 2 | 237 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
