Assay Details
Gene Symbol: ccndx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016108.2 | NP_001016108.1 | 3 - 4 | 616 | 72 | |
GenBank mRNA | BC158907.1 | - | 3 - 4 | 619 | 72 | |
CR762119.2 | - | 3 - 4 | 616 | 72 | ||
DQ508541.1 | - | 3 - 4 | 616 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
