Assay Details
Gene Symbol: alg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016120.2 | NP_001016120.1 | 2 - 3 | 505 | 88 | |
GenBank mRNA | BC121879.1 | - | 2 - 3 | 786 | 88 | |
BC155512.1 | - | 2 - 3 | 747 | 88 | ||
CR762085.2 | - | 2 - 3 | 505 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
