Assay Details
Gene Symbol: aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016129.2 | NP_001016129.1 | 10 - 11 | 1401 | 91 | |
XM_012962024.2 | XP_012817478.1 | 10 - 11 | 1554 | 91 | ||
GenBank mRNA | BC161161.1 | - | 10 - 11 | 1442 | 91 | |
BC170898.1 | - | 10 - 11 | 1370 | 91 | ||
BC170900.1 | - | 10 - 11 | 1370 | 91 | ||
CR762057.2 | - | 10 - 11 | 1401 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
