Assay Details
| Gene Symbol: capns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016161.2 | NP_001016161.1 | 1 - 2 | 139 | 72 | |
| GenBank mRNA | BC170827.1 | - | 1 - 2 | 134 | 72 | |
| BC170831.1 | - | 1 - 2 | 134 | 72 | ||
| CR761923.2 | - | 1 - 2 | 139 | 72 | ||
More Information
Set Membership: |
|

capns2
Genomic Map