Assay Details
Gene Symbol: capns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016161.2 | NP_001016161.1 | 9 - 10 | 705 | 87 | |
GenBank mRNA | BC170827.1 | - | 9 - 10 | 700 | 87 | |
BC170831.1 | - | 9 - 10 | 700 | 87 | ||
CR761923.2 | - | 9 - 10 | 705 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
