Assay Details
Gene Symbol: hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016165.2 | NP_001016165.1 | 1 - 2 | 119 | 67 | |
GenBank mRNA | BC158373.1 | - | 1 - 2 | 109 | 67 | |
CR761909.2 | - | 1 - 2 | 119 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
