Assay Details
| Gene Symbol: cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016170.2 | NP_001016170.1 | 1 - 2 | 80 | 68 | |
| GenBank mRNA | BC160465.1 | - | 1 - 2 | 140 | 68 | |
| BC170572.1 | - | 1 - 2 | 79 | 68 | ||
| BC170574.1 | - | 1 - 2 | 79 | 68 | ||
| CR761887.2 | - | 1 - 2 | 80 | 68 | ||
More Information
Set Membership: |
|

cks2
Genomic Map