Assay Details
| Gene Symbol: adprhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016184.2 | NP_001016184.1 | 3 - 4 | 469 | 71 | |
| GenBank mRNA | CR761827.2 | - | 3 - 4 | 469 | 71 | |
More Information
Set Membership: |
|

adprhl2
Genomic Map