Assay Details
Gene Symbol: ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016242.2 | NP_001016242.1 | 386 | 63 | ||
GenBank mRNA | BC167327.1 | - | 383 | 63 | ||
CR761787.2 | - | 386 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
