Assay Details
| Gene Symbol: mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016249.2 | NP_001016249.1 | 2 - 3 | 250 | 77 | |
| GenBank mRNA | BC135459.1 | - | 2 - 3 | 214 | 77 | |
| CR761757.2 | - | 2 - 3 | 250 | 77 | ||
More Information
Set Membership: |
|

mrpl40
Genomic Map