Assay Details
Gene Symbol: popdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016293.2 | NP_001016293.1 | 3 - 4 | 982 | 75 | |
XM_018093881.1 | XP_017949370.1 | 4 - 5 | 1233 | 75 | ||
GenBank mRNA | BC160588.1 | - | 3 - 4 | 1033 | 75 | |
BC171320.1 | - | 2 - 3 | 919 | 75 | ||
BC171321.1 | - | 2 - 3 | 919 | 75 | ||
CR761552.2 | - | 3 - 4 | 982 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
