Assay Details
| Gene Symbol: casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016299.2 | NP_001016299.1 | 1 - 2 | 317 | 79 | |
| GenBank mRNA | BC170775.1 | - | 1 - 2 | 315 | 79 | |
| BC170801.1 | - | 1 - 2 | 315 | 79 | ||
| CR761526.2 | - | 1 - 2 | 317 | 79 | ||
More Information
Set Membership: |
|

casp7
Genomic Map