Assay Details
| Gene Symbol: ppp1r14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016307.2 | NP_001016307.1 | 2 - 3 | 296 | 71 | |
| GenBank mRNA | CR761497.2 | - | 2 - 3 | 296 | 71 | |
More Information
Set Membership: |
|

ppp1r14b
Genomic Map