Assay Details
Gene Symbol: cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016333.2 | NP_001016333.1 | 2 - 3 | 848 | 84 | |
XM_012962186.2 | XP_012817640.1 | 3 - 4 | 690 | 84 | ||
XM_012962189.2 | XP_012817643.1 | 3 - 4 | 689 | 84 | ||
XM_012962191.1 | XP_012817645.1 | 3 - 4 | 606 | 84 | ||
XM_018089476.1 | XP_017944965.1 | 2 - 3 | 356 | 84 | ||
GenBank mRNA | BC171112.1 | - | 2 - 3 | 760 | 84 | |
CR761413.2 | - | 2 - 3 | 848 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
