Assay Details
| Gene Symbol: psenen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016336.2 | NP_001016336.1 | 2 - 3 | 333 | 87 | |
| XM_012966572.2 | XP_012822026.1 | 2 - 3 | 259 | 87 | ||
| GenBank mRNA | BC121544.1 | - | 2 - 3 | 295 | 87 | |
| CR761406.2 | - | 2 - 3 | 333 | 87 | ||
More Information
Set Membership: |
|

psenen
Genomic Map