Assay Details
Gene Symbol: fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016344.2 | NP_001016344.1 | 8 - 9 | 1374 | 81 | |
XM_018093883.1 | XP_017949372.1 | 8 - 9 | 1584 | 81 | ||
XM_018093884.1 | XP_017949373.1 | 7 - 8 | 1500 | 81 | ||
GenBank mRNA | CR761373.2 | - | 8 - 9 | 1374 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
