Assay Details
Gene Symbol: nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016355.2 | NP_001016355.1 | 5 - 6 | 760 | 62 | |
XM_012970412.2 | XP_012825866.1 | 6 - 7 | 1213 | 62 | ||
XM_012970413.2 | XP_012825867.1 | 6 - 7 | 1136 | 62 | ||
XM_012970414.2 | XP_012825868.1 | 6 - 7 | 1042 | 62 | ||
GenBank mRNA | BC135188.1 | - | 6 - 7 | 1047 | 62 | |
CR761311.2 | - | 5 - 6 | 760 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
